Alfa Aesar™ Dodecacarbonyltetrairidium, 98%

Manufacturer: Alfa Aesar™ 1050503

Quantity 1g

 View more versions of this product

Catalog No. AA1050503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 11065-24-0
    Formula Weight 1105.01
    MDL Number MFCD00011064
    UN Number UN3466
    Synonym Tetrairidium dodecacarbonyl; Iridium carbonyl
    Chemical Name or Material Dodecacarbonyltetrairidium

    Specifications

    Formula Weight 1105.01
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN3466
    Melting Point 195°C (dec.)
    Health Hazard 2 GHS H Statement
    H300-H330
    Fatal if swallowed.
    Fatal if inhaled.
    MDL Number MFCD00011064
    CAS 11065-24-0
    Synonym Tetrairidium dodecacarbonyl; Iridium carbonyl
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
    Sensitivity Air Sensitive
    Quantity 1g
    Physical Form Powder
    Health Hazard 1 Danger
    Chemical Name or Material Dodecacarbonyltetrairidium
    Assay 98%

    GHS H Statement
    H300-H330
    Fatal if swallowed.
    Fatal if inhaled.

    GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit