Alfa Aesar™ Dodecacarbonyltriruthenium, 99%

Carbonylation

Manufacturer: Alfa Aesar™ 1050703

Quantity 1g

 View more versions of this product

Catalog No. AA1050703

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15243-33-1
    Linear Formula Ru3(CO)12
    Formula Weight 639.33
    MDL Number MFCD00011209
    UN Number UN3466
    Synonym Ruthenium carbonyl; Triruthenium dodecacarbonyl
    Chemical Name or Material Dodecacarbonyltriruthenium

    Specifications

    Formula Weight 639.33
    Health Hazard 3 GHS P Statement
    P261-P304+P340-P311-P405-P403+P233-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    Store locked up.
    Store in a well-ventilated place.
    Keep container tightly closed.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    UN Number UN3466
    Melting Point ≈150°C (dec.)
    CAS 15243-33-1
    Solubility Information Sparingly soluble in hydrocarbons (e.g. hexane, cyclohexane, benzene) and acetone; Solutions undergo some decomposition on strong heating
    Health Hazard 1 Danger
    Physical Form Crystalline
    Linear Formula Ru3(CO)12
    MDL Number MFCD00011209
    Synonym Ruthenium carbonyl; Triruthenium dodecacarbonyl
    Color Orange
    Health Hazard 2 GHS H Statement
    H331
    Toxic if inhaled.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
    Quantity 1g
    Chemical Name or Material Dodecacarbonyltriruthenium
    Assay 99%

    GHS H Statement
    H331
    Toxic if inhaled.

    GHS P Statement
    P261-P304+P340-P311-P405-P403+P233-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    Store locked up.
    Store in a well-ventilated place.
    Keep container tightly closed.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit