Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecane, ≥99.8% (GC), MilliporeSigma™ Supelco™

Dodecane is a clear, colorless liquid, having a mild aliphatic hydrocarbon odor, which belongs to the family of acyclic alkanes and contains compounds like 3-methylundecane and 5-methylundecane.

$64.60 - $251.00

Specifications

Percent Purity ≥99.8% (GC)
Density 0.75 g/mL (at 25°C)
Grade Analytical Standard
Linear Formula CH3(CH2)10 CH3
Physical Form Neat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-9799
View Documents
MilliporeSigma Supelco
440105ML
5 mL
Each for $64.60
Add to cart
 
11-101-8375
View Documents
MilliporeSigma Supelco
4401025ML
25 mL
Each for $251.00
Add to cart
 
Description

Description

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications

Specifications

≥99.8% (GC)
Analytical Standard
Neat
0.75 g/mL (at 25°C)
CH3(CH2)10 CH3
Safety and Handling

Safety and Handling

ShelfLife : Limited shelf life, expiry date on the label

RTECSNumber : JR2125000

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit