Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Dodecane, 99%, pure, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
250mL
500mL
2.5L
10L
Packaging:
Glass bottle
Metal Drum
112-40-3
C12H26
170.34
MFCD00008969
SNRUBQQJIBEYMU-UHFFFAOYSA-N
n-dodecane,dihexyl,bihexyl,adakane 12,n-dodecan,n-dodecan german,duodecane,alkanes, c10-14,undecane, methyl,ccris 661
8182
CHEBI:28817
dodecane
CCCCCCCCCCCC
112-40-3
C12H26
170.34
MFCD00008969
SNRUBQQJIBEYMU-UHFFFAOYSA-N
n-dodecane,dihexyl,bihexyl,adakane 12,n-dodecan,n-dodecan german,duodecane,alkanes, c10-14,undecane, methyl,ccris 661
8182
CHEBI:28817
dodecane
CCCCCCCCCCCC
Specific Gravity 0.753
Viscosity <7 mm2/s (40°C)
Vapor Pressure 1mbar at 48°C
Refractive Index 1.4210 to 1.4230
Packaging Glass bottle
Density 0.7530g/mL
Infrared Spectrum Authentic
Flash Point 71°C
Color Undesignated
Quantity 2.5L
Melting Point -12.0°C
Boiling Point 215.0°C to 217.0°C
Assay Percent Range 99%
Linear Formula CH3(CH2)10CH3
Beilstein 01, 171
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone and ether
Formula Weight 170.34
Physical Form Liquid
Percent Purity 99%
Grade Pure
Chemical Name or Material n-Dodecane, 99%
Show More Show Less

GHS P Statement
May be fatal if swallowed and enters airways.
Repeated exposure may cause skin dryness or cracking.

Emergency Overview
Combustible liquid. Harmful by inhalation. Aspiration hazard if swallowed - can enter lungs and cause damage. Repeated exposure may cause skin dryness or cracking.
NFPA

Danger

EINECSNumber : 203-967-9

RTECSNumber : JR2125000

TSCA : TSCA