Learn More
Dodecanedioic acid, 99%, Thermo Scientific Chemicals
Description
Dodecanedioic acid is used as an intermediate for plasticizers, lubricants and adhesives. It is also used in antiseptics, top-grade coatings, painting materials, corrosion inhibitor and surfactant. It finds application in engineering plastics such as nylon 612.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Dodecanedioic acid is used as an intermediate for plasticizers, lubricants and adhesives. It is also used in antiseptics, top-grade coatings, painting materials, corrosion inhibitor and surfactant. It finds application in engineering plastics such as nylon 612.
Solubility
Soluble in water, ethanol, hot methanol, hot toluene and hot acetic acid.
Notes
Incompatible with strong oxidizing agents and reducing agents.
Specifications
Melting Point | 127°C to 131°C |
Density | 1.15 |
Boiling Point | 244°C to 245°C (10 mmHg) |
Flash Point | 227°C (440°F) |
Odor | Odorless |
Quantity | 250 g |
Beilstein | 1782580 |
Solubility Information | Soluble in water,ethanol,hot methanol,hot toluene and hot acetic acid. |
Formula Weight | 230.31 |
Percent Purity | 99% |
Show More |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H319 Causes serious eye irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 211-746-3 |
RTECSNumber | JR2370000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.