Alfa Aesar™ Dodecanedioic acid, 99%

Manufacturer: Alfa Aesar™ A103870B

Quantity 1000g

 View more versions of this product

Catalog No. AAA103870B

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 693-23-2
    Molecular Formula HO2C(CH2)10CO2H
    Formula Weight 230.31
    MDL Number MFCD00002735
    Beilstein 1782580
    Synonym 1,10-Decanedicarboxylic acid
    Chemical Name or Material Dodecanedioic acid

    Specifications

    Formula Weight 230.31
    Health Hazard 3 GHS P Statement
    P280-P264-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.
    Molecular Formula HO2C(CH2)10CO2H
    CAS 693-23-2
    Health Hazard 1 Warning
    Odor Odorless
    TSCA TSCA
    Synonym 1,10-Decanedicarboxylic acid
    Flash Point 227°C (440°F)
    Quantity 1000g
    MDL Number MFCD00002735
    Beilstein 1782580
    Boiling Point 244° to 245°C (10mmHg)
    Melting Point 127° to 131°C
    Health Hazard 2 GHS H Statement
    H319
    Causes serious eye irritation.
    RTECS Number JR2370000
    Chemical Name or Material Dodecanedioic acid
    Assay 99%

    Description

    Applications
    Dodecanedioic acid is used as an intermediate for plasticizers, lubricants and adhesives. It is also used in antiseptics, top-grade coatings, painting materials, corrosion inhibitor and surfactant. It finds application in engineering plastics such as nylon 612.

    Solubility
    Soluble in water, ethanol, hot methanol, hot toluene and hot acetic acid.

    Notes
    Incompatible with strong oxidizing agents and reducing agents.

    GHS H Statement
    H319
    Causes serious eye irritation.

    GHS P Statement
    P280-P264-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit