Alfa Aesar™ Dodecylbenzene, mixture of isomers

Manufacturer: Alfa Aesar™ 31362A1

Quantity 1kg

 View more versions of this product

Catalog No. AA31362A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 123-01-3
    Formula Weight 246.44
    MDL Number MFCD00008974
    Synonym 1-Phenyldodecane
    Chemical Name or Material Dodecylbenzene, Mixture of isomers

    Specifications

    Formula Weight 246.44
    Density 0.87g/mL
    MDL Number MFCD00008974
    Synonym 1-Phenyldodecane
    Melting Point −3°C
    Odor Specific type
    TSCA TSCA
    Physical Form Liquid
    CAS 123-01-3
    Refractive Index 1.49
    Boiling Point 179° to 180°C (13mmHg)
    Quantity 1kg
    Chemical Name or Material Dodecylbenzene, Mixture of isomers

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit