Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecyldimethylchlorosilane, 97%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
Packaging:
Glass bottle
66604-31-7
C14H31ClSi
262.93
DLLABNOCKQMTEJ-UHFFFAOYSA-N
dodecyldimethylchlorosilane,chloro dodecyl dimethylsilane,1-chlorododecyl-dimethyl-silicon,chlorododecyldimethylsilane,acmc-209nvw,dimethyldodecylchlorosilane,dimethyldodecylsilyl chloride,n-dodecyldimethylchlorosilane,dimethyl-dodecyl-chlorosilane,chloro-dimethyl-dodecylsilane
2758731
chloro-dodecyl-dimethylsilane
CCCCCCCCCCCC[Si](C)(C)Cl
66604-31-7
C14H31ClSi
262.93
DLLABNOCKQMTEJ-UHFFFAOYSA-N
dodecyldimethylchlorosilane,chloro dodecyl dimethylsilane,1-chlorododecyl-dimethyl-silicon,chlorododecyldimethylsilane,acmc-209nvw,dimethyldodecylchlorosilane,dimethyldodecylsilyl chloride,n-dodecyldimethylchlorosilane,dimethyl-dodecyl-chlorosilane,chloro-dimethyl-dodecylsilane
2758731
chloro-dodecyl-dimethylsilane
CCCCCCCCCCCC[Si](C)(C)Cl
Density 0.8650g/mL
Specific Gravity 0.865
Refractive Index 1.4440 to 1.4460 (20°C, 589nm)
Quantity 100mL
Boiling Point 291.0°C to 293.0°C
Infrared Spectrum Authentic
Flash Point 110°C
Color Colorless
Packaging Glass bottle
Assay Percent Range 96% min. (GC)
Linear Formula Cl(CH3)2Si(CH2)11CH3
Solubility Information Solubility in water: insoluble.
Formula Weight 262.93
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Dodecyldimethylchlorosilane
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 266-421-9