Learn More
Selleck Chemical LLC
Dofetilide 50mg 115256-11-6 UK-68798
Manufacturer: Selleck Chemical LLC S165850MG
Dofetilide (UK-68798) is a selective potassium channel ((hERG)) blocker, used as a Class III antiarrhythmic drug.