Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DOG1/TMEM16A Mouse anti-Human, PE/Cy5.5, Clone: DG1/1484, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254492PECY55

Catalog No. N254492PEC5


Add to cart

Description

Description

DOG1/TMEM16A Monoclonal antibody specifically detects DOG1/TMEM16A in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

DOG1/TMEM16A
DG1/1484
Flow Cytometry
Conjugated, Purified
ANO1
Recombinant human DOG-1 protein fragment (aa 2-101) (exact sequence is proprietary)
Protein A or G purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
ANO1, anoctamin 1, DOG1, ORAOV2, TAOS2, TMEM16A
Mouse
IgG2b Kappa
0.1 mL
Primary
55107.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit