Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DOG1/TMEM16A Rabbit anti-Human, Clone: DG1/1487R, Novus Biologicals™

Rabbit Monoclonal Antibody

$211.22 - $435.85

Specifications

Antigen DOG1/TMEM16A
Clone DG1/1487R
Host Species Rabbit
Isotype IgG Kappa
Gene Alias ANO1, anoctamin 1, DOG1, ORAOV2, TAOS2, TMEM16A
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB25313720 View Documents Novus Biologicals
NBP25313720UG
20 ug Each for $211.22
Add to cart
 
NB253137100 View Documents Novus Biologicals
NBP253137100UG
100 ug Each for $435.85
Add to cart
 
NB253137200 View Documents Novus Biologicals
NBP253137200UG
200 ug This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

DOG1/TMEM16A Monoclonal antibody specifically detects DOG1/TMEM16A in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

DOG1/TMEM16A
Rabbit
ANO1, anoctamin 1, DOG1, ORAOV2, TAOS2, TMEM16A
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Store at 4C.
Monoclonal
0.2 mg/mL
DG1/1487R
IgG Kappa
Human
RUO
Purified
Primary
10mM PBS with 0.05% BSA with 0.05% Sodium Azide

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit