Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DOG1/TMEM16A Rabbit anti-Human, Clone: DG1/2564R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen DOG1/TMEM16A
Clone DG1/2564R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122777
View Documents
Novus Biologicals
NBP30765210UG
100 μg
Each for $406.52
Add to cart
 
NB122778
View Documents
Novus Biologicals
NBP30765220UG
20 μg
Each for $159.82
Add to cart
 
Description

Description

DOG1/TMEM16A Monoclonal antibody specifically detects DOG1/TMEM16A in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

DOG1/TMEM16A
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
10 mM PBS with 0.05% BSA
55107
IgG
Primary
Store at 4°C.
DG1/2564R
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Rabbit
Human
ANO1, anoctamin 1, DOG1, ORAOV2, TAOS2, TMEM16A
A recombinant human DOG1/TMEM16A protein fragment (around aa 2-101) (exact sequence is proprietary)
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit