Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dogfish Sharks

Squalus acanthius.
Supplier: Ranaco Corporation 132.1135.10
Description
Sharks and the cartilaginous skeletoned fishes are representatives of a group of fish whose ancestors were the first jaw-bearing vertebrates. Except for skeletal degeneration, sharks are good examples of a fairly primitive type.
- These sharks are from the Class Chondrichthyes Elasmobranchii squalus, the dogfish sharks
- Shark color injections differ from the standard color progression (see Injection Key)
- Packed in Bio-Fresh™
Order Info
INJECTION KEY
- Plain: Circulatory system not injected with colors
- Single: Arterial system injected with red latex
- Double: Hepatic portal system injected with blue latex
- Triple: Systemic veins injected with yellow latex
Specifications
Dogfish Shark | |
Double | |
55.88 to 68.58 cm |
Bio-Fresh™ | |
22 to 27 in. | |
10 Pk. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title