Learn More
Dogfish Sharks
Squalus acanthius.
Supplier: Ranaco Corporation 132.1055.1
Description
Sharks and the cartilaginous skeletoned fishes are representatives of a group of fish whose ancestors were the first jaw-bearing vertebrates. Except for skeletal degeneration, sharks are good examples of a fairly primitive type.
- These sharks are from the Class Chondrichthyes Elasmobranchii squalus, the dogfish sharks
- Shark color injections differ from the standard color progression (see Injection Key)
- Packed in Bio-Fresh™
Order Info
INJECTION KEY
- Plain: Circulatory system not injected with colors
- Single: Arterial system injected with red latex
- Double: Hepatic portal system injected with blue latex
- Triple: Systemic veins injected with yellow latex
Specifications
Dogfish Shark | |
Double | |
68.58+ cm |
Bio-Fresh™ | |
27+ in. | |
Ea. |