Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dogfish Sharks
SCI_ED

Squalus acanthius.

 View more versions of this product

Catalog No. S1508


Due to product restrictions, please to purchase or view availability for this product.


Description

Description

Sharks and the cartilaginous skeletoned fishes are representatives of a group of fish whose ancestors were the first jaw-bearing vertebrates. Except for skeletal degeneration, sharks are good examples of a fairly primitive type.

  • These sharks are from the Class Chondrichthyes Elasmobranchii squalus, the dogfish sharks
  • Shark color injections differ from the standard color progression (see Injection Key)
  • Packed in Bio-Fresh™

Order Info

INJECTION KEY

  • Plain: Circulatory system not injected with colors
  • Single: Arterial system injected with red latex
  • Double: Hepatic portal system injected with blue latex
  • Triple: Systemic veins injected with yellow latex

Specifications

Specifications

Dogfish Shark
Triple
55.88 to 68.58 cm
Bio-Fresh™
22 to 27 in.
12/Cs.
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit