Learn More
Dogfish Sharks
SCI_ED
Squalus acanthius.
$35.00 - $248.00
Specifications
Preservative Bio-Fresh™
View More Specs
Products 12
Catalog Number Mfr. No. Injection Type Length (English) Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Injection Type Length (English) Type Quantity Price Quantity & Availability  
S1030S
SCI_ED
View Documents
Ranaco Corporation
132.1035.1
Single 27+ in. Dogfish Shark Ea.
Pack for $27.25
N/A
Due to product restrictions, please to purchase or view availability for this product.
S1501
SCI_ED
View Documents
Ranaco Corporation
132.1095.1
Plain 22 to 27 in. Dogfish Shark Ea.
Pack for $19.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
S1502
SCI_ED
View Documents
Ranaco Corporation
132.1135.1
Double 22 to 27 in. Dogfish Shark 12/Cs.
Pack for $27.75
N/A
Due to product restrictions, please to purchase or view availability for this product.
S1504
SCI_ED
View Documents
Ranaco Corporation
132.1055.1
Double 27+ in. Dogfish Shark Ea.
Pack for $27.75
N/A
Due to product restrictions, please to purchase or view availability for this product.
S1503S
SCI_ED
View Documents
Ranaco Corporation
132.1015.1
Plain 27+ in. Dogfish Shark 10/Cs.
Pack for $25.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
S1505
SCI_ED
View Documents
Ranaco Corporation
132.1075.1
Triple 27+ in. Dogfish Shark 1/Pk.
Pack for $35.00
Add to cart
S1508
SCI_ED
View Documents
Ranaco Corporation
132.1155.1
Triple 22 to 27 in. Dogfish Shark 12/Cs.
Pack for $28.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
S1509S
SCI_ED
View Documents
Ranaco Corporation
132.1175.1
Plain 18 to 22 in. Dogfish Shark 15/Cs.
Pack for $13.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
S07053
SCI_ED
View Documents
Ranaco Corporation
132.1135.10
Double 22 to 27 in. Dogfish Shark 10 Pk.
Pack of 10 for $248.00
Add to cart
S1207S
SCI_ED
View Documents
N/A Plain Dogfish Shark Pup 10 Pk. Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
S1510S
SCI_ED
View Documents
N/A Double 18 to 22 in. Dogfish Shark 15/Cs. Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
S1511S
SCI_ED
View Documents
N/A Plain 18 to 22 in. Dogfish Shark Pup Ea. Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
Sharks and the cartilaginous skeletoned fishes are representatives of a group of fish whose ancestors were the first jaw-bearing vertebrates. Except for skeletal degeneration, sharks are good examples of a fairly primitive type.
  • These sharks are from the Class Chondrichthyes Elasmobranchii squalus, the dogfish sharks
  • Shark color injections differ from the standard color progression (see Injection Key)
  • Packed in Bio-Fresh™
Order Info
INJECTION KEY
  • Plain: Circulatory system not injected with colors
  • Single: Arterial system injected with red latex
  • Double: Hepatic portal system injected with blue latex
  • Triple: Systemic veins injected with yellow latex
Specifications
Specifications
Bio-Fresh™