Learn More
Abnova™ DOHH 293T Cell Transient Overexpression Lysate (Denatured) (T03)
Human overexpression lysate
Manufacturer:  Abnova™ H00083475T03
Catalog No. 89-942-311
Add to cart
Specifications
Specifications
Western Blot
293T
NM_031304
pCMV-DOHH full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
DOHH 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 32.9kDa
Transient overexpression cell lysate was tested with anti-DOHH antibody (Mfr. No. H00083475-B01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)