Learn More
Invitrogen™ DOHH Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114169
Description
Immunogen sequence: GQMQDARAIP MLVDVLQDTR QEPMVRHEAG EALGAIGDPE VLEILKQYSS For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 92%; Guinea Pig: 77%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 92%; Rat: 93%; Yeast: 100%; Zebrafish: 92%.

Specifications
DOHH | |
Polyclonal | |
Unconjugated | |
DOHH | |
Deoxyhypusine dioxygenase; deoxyhypusine hydroxylase; deoxyhypusine hydroxylase/monooxygenase; deoxyhypusine monooxygenase; DOHH; hDOHH; HEAT-like (PBS lyase) repeat containing 1; HEAT-like repeat-containing protein 1; HLRC1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
83475 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BU89 | |
DOHH | |
Immunogen sequence: GQMQDARAIP MLVDVLQDTR QEPMVRHEAG EALGAIGDPE VLEILKQYSS (aa 63-112). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.