Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DOK2 Mouse anti-Human, PE/Atto594, Clone: OTI1A4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271407PEATT594

Catalog No. N271407P594


Add to cart

Description

Description

DOK2 Monoclonal antibody specifically detects DOK2 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

DOK2
OTI1A4
Flow Cytometry
Affinity Purified, Conjugated
DOK2
Full length recombinant protein of human DOK2 (NP_003965) produced in HEK293T cells.
Immunogen affinity purified
Phospho Specific, Signal Transduction
Primary
9046.0
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
docking protein 2, docking protein 2, 56kD, docking protein 2, 56kDa, Dok-2, Downstream of tyrosine kinase 2, p56(dok-2), P56DOK, p56dok-2
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit