Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DOK7 Mouse anti-Human, Clone: OTI2C12, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA504787
Description
The protein encoded by this gene is essential for neuromuscular synaptogenesis. The protein functions in aneural activation of muscle-specific receptor kinase, which is required for postsynaptic differentiation, and in the subsequent clustering of the acetylcholine receptor in myotubes. This protein can also induce autophosphorylation of muscle-specific receptor kinase. Mutations in this gene are a cause of familial limb-girdle myasthenia autosomal recessive, which is also known as congenital myasthenic syndrome type 1B. Alternative splicing results in multiple transcript variants.Specifications
DOK7 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q18PE1 | |
C4orf25, CMS10, CMS1B | |
Mouse | |
IgG2b | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
285489 |
Western Blot | |
OTI2C12 | |
Unconjugated | |
DOK7 | |
Liquid | |
DOK7 | |
Full length human recombit protein of human DOK7 produced in HEK293T cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title