Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Dolasetron mesylate

Potent and selective 5-HT3 antagonist

$69.00 - $289.00

Specifications

Chemical Name or Material 3-Oxooctahydro-2,6-methano-2H-quinolizin-8-yl 1H-indole-3-carboxylate methanesulfonate
Molecular Formula C19H20N2O3.CH3SO3H
CAS 115956-13-3
Product Type Target: 5-HT3 Receptor Antagonists
Formula Weight 420.48
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
562910 View Documents Tocris Bioscience™
5629/10
10mg Each for $69.00
Add to cart
 
562950 View Documents Tocris Bioscience™
5629/50
50mg Each for $289.00
Add to cart
 
Description

Description

Potent and selective 5-HT3 receptor antagonist (pIC50 = 7.55). Exhibits 200-fold selectivity for 5-HT3 over 5-HT1A, 5-HT1B and 5-HT2 receptors, and a range of other GPCRs. Blocks the 5-HT-elicited von Bezold-Jarisch reflex in rats and suppresses cisplatin -induced vomiting in ferrets.
Specifications

Specifications

3-Oxooctahydro-2,6-methano-2H-quinolizin-8-yl 1H-indole-3-carboxylate methanesulfonate
115956-13-3
420.48
C19H20N2O3.CH3SO3H
Target: 5-HT3 Receptor Antagonists
Safety and Handling

Safety and Handling

Recommended Storage : Store at 4°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit