Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DOLK Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549645

Catalog No. PA549645

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Involved in the synthesis of the sugar donor Dol-P-Man which is required in the synthesis of N-linked and O-linked oligosaccharides and for that of GPI anchors.
Specifications

Specifications

DOLK
Polyclonal
PBS with 0.09% sodium azide
Q9UPQ8
Dolichol kinase, Transmembrane protein 15, DOLK, KIAA1094, TMEM15
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
22845
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
DOLK
Liquid
DOLK
KLH conjugated synthetic peptide between 312-339 amino acids from the Central region of human DOLK.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit