Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DOLPP1 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548614

Catalog No. PA548614

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Required for efficient N-glycosylation. Necessary for maintaining optimal levels of dolichol-linked oligosaccharides. Hydrolyzes dolichyl pyrophosphate at a very high rate and dolichyl monophosphate at a much lower rate. Does not act on phosphatidate.
Specifications

Specifications

DOLPP1
Polyclonal
PBS with 0.09% sodium azide
Q9JMF7
Dolichyldiphosphatase 1, Dolichyl pyrophosphate phosphatase 1, DOLPP1, LSFR2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
57170
Western Blot
Unconjugated
Dolpp1
Liquid
DOLPP1
KLH conjugated synthetic peptide between 207-236 amino acids from the C-terminal region of human DOLPP1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit