Learn More
domain-I-of-FIZ-1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 158261AP
Catalog No. 50-173-0511
Add to cart
Description
Description
FLJ14768 is homologous to mouse and bovine Fiz1 (flt-3 interacting zinc finger). It interacts with NRL (neural retina leucine zipper). Possible role for Fiz1 as a transcriptional repressor.
Specifications
Specifications
domain-I-of-FIZ-1
0.44 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q96SL8, Q9WTJ4
domain I of FIZ 1, domain II of FIZ1, FIZ1, FLT3 interacting zinc finger 1, Zinc finger protein 798, ZNF798
Rabbit
IgG
150 μL
-20°C
Primary
23877, 292584, 84922
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
FIZ1
Liquid
FIZ1
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat