Learn More
MCR Safety Dominator™ 3
Click to view available options
Lens Tint:
Amber
Blue
Clear
Gray
Indoor/Outdoor
Mirror
Frame Color:
Black
Blue, Yellow
Clear
Gun Metal
Lens Coating:
Anti-fog
Scratch-resistant
Description
- Italian fashion, sport focused design
- Co-injected wire core independently adjustable nose pads
- Co-injected soft wire core adjustable TPR temples
- MAX3™, MAX6™, or BOSSMAN™ Mirror coatings
- Meets or Exceeds Z87+ & Military High Velocity
- Gun Metal/Red TPR, Clear/Blue TPR, Black/Black TPR, or Yellow/Blue TPR
Certifications
CSA
Compliance
Meets ANSI Z87.1-2015, CSA Z94.3 and MIL - PRF - 31013 standards.
Specifications
Lens Tint | Clear |
For Use With (Application) | Manufacturing |
Frame Color | Blue, Yellow |
No. per Pack | 1 Pair |
Lens Material | Polycarbonate |
Frame Material | Polycarbonate |