Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCR Safety Dominator PVC/Polyester Rain Jacket
Click to view available options
Size:
2X-Large
3X-Large
Large
Medium
X-Large
Description
- Jacket with removable drawstring hood, take up snaps at wrists
- PVC/Hi-tensil polyester/PVC
- Zipper front with double storm flap, no pockets or vents
- Inner sleeves to prevent splash back
- Limited Flammability
- NOT intended for use as protection against ARC Flash or Flash Fire
Specifications
Size | Medium |
Gender | Unisex |
Material | Polyester, PVC |
Color | Green |
Closure Type | Zipper |
Certifications/Compliance | Limited Flammability |
Cuff Style | Snap |
Disposable | No |
Finish | Smooth |
Hood Type | Detachable |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction