Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Domoic acid, Tocris Bioscience™

Click to view available options
Quantity:
1mg
5mg

Chemical Identifiers

14277-97-5
C15H21NO6
311.334
VZFRNCSOCOPNDB-AOKDLOFSSA-N
(2S,3S,4R,5'R)-2-Carboxy-4-(5'-carboxy-1'-methyl-1Z,3E-hexadienyl)-3-pyrrolidineacetic acid
5282253
CHEBI:34727
Quantity 5mg
CAS 14277-97-5
Molecular Formula C15H21NO6
Synonym (2S,3S,4R,5'R)-2-Carboxy-4-(5'-carboxy-1'-methyl-1Z,3E-hexadienyl)-3-pyrrolidineacetic acid
InChI Key VZFRNCSOCOPNDB-AOKDLOFSSA-N
SMILES CC(C=CC=C(C)C1CNC(C1CC(=O)O)C(=O)O)C(=O)O
IUPAC Name (2S,3S,4S)-4-[(2Z,4E,6R)-6-carboxyhepta-2,4-dien-2-yl]-3-(carboxymethyl)pyrrolidine-2-carboxylic acid
Molecular Weight (g/mol) 311.334
PubChem CID 5282253
ChEBI CHEBI:34727
Formula Weight 311.33
Chemical Name or Material Domoic acid
Grade >95%
Show More Show Less

Recommended Storage : Desiccate at -20°C