Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Domperidone, Tocris Bioscience™

Catalog Number 253650
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
57808-66-9
C22H24ClN5O2
425.917
FGXWKSZFVQUSTL-UHFFFAOYSA-N
domperidone, motilium, nauzelin, domperidonum, domperidona, domperidon, domperidonum inn-latin, domperidona inn-spanish, motillium, motinorm
3151
CHEBI:31515
6-chloro-3-[1-[3-(2-oxo-3H-benzimidazol-1-yl)propyl]piperidin-4-yl]-1H-benzimidazol-2-one
C1CN(CCC1N2C3=C(C=C(C=C3)Cl)NC2=O)CCCN4C5=CC=CC=C5NC4=O
This item is not returnable. View return policy
57808-66-9
C22H24ClN5O2
425.917
FGXWKSZFVQUSTL-UHFFFAOYSA-N
domperidone, motilium, nauzelin, domperidonum, domperidona, domperidon, domperidonum inn-latin, domperidona inn-spanish, motillium, motinorm
3151
CHEBI:31515
6-chloro-3-[1-[3-(2-oxo-3H-benzimidazol-1-yl)propyl]piperidin-4-yl]-1H-benzimidazol-2-one
C1CN(CCC1N2C3=C(C=C(C=C3)Cl)NC2=O)CCCN4C5=CC=CC=C5NC4=O

Peripheral D2-like antagonist

Peripheral dopamine D2-like receptor antagonist that does not readily cross the blood brain barrier. Displays gastroprokinetic and antiemetic properties; increases the frequency and duration of antral and duodenal contractions and protects from apomorphine-induced emesis (ED50 values are 0.003 and 0.03 mg/kg for i.v. and oral administration respectively).
Quantity 50 mg
Formula Weight 425.92
Percent Purity >99%
Chemical Name or Material Domperidone
Recommended Storage Store at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit