Learn More
Donkey anti-Human IgG whole molecule, HRP, Polyclonal, Secondary Antibody, Abnova™
Manufacturer:  Abnova Corporation PAB10686
Catalog No. 89-137-226
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
Donkey anti-Human IgG whole molecule
HRP
The optimal working dilution should be determined by the end user.
Lyophilized
Human IgG whole molecule
2 mg
Secondary
Human
Dot Blot, ELISA, Immunohistochemistry, Western Blot
Donkey anti-human IgG recognizes human IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Peroxidase.
Lyophilized from 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% gentamicLyophilized from sulfate)
Donkey
This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities.
Store at 4°C on dry atmosphere. After reconstitution with 1mL of deionized water, store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal