Donkey anti-Mouse IgG, Species Adsorbed: Hu, Rt, B, H, G, O, Rb, Ck, Gp, Secondary Antibody Millipore™ (Chemicon™)

Manufacturer: EMD Millipore AP192B/DEL

 View more versions of this product

Catalog No. 50-174-489

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Antigen Donkey anti-Mouse IgG, Species Adsorbed: Hu, Rt, B, H, G, O, Rb, Ck, Gp
    Format Affinity Purified
    Host Species Donkey
    Quantity 500μL
    Regulatory Status RUO
    Research Discipline Enzyme
    Primary or Secondary Secondary
    Monoclonal or Polyclonal Polyclonal
    Conjugate Biotin
    Cross Reactivity Mouse

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit