Learn More
Donkey anti-Rabbit IgG whole molecule,Polyclonal Secondary Antibody, Abnova™
Manufacturer:  Abnova Corporation PAB9381
Catalog No. 89-139-044
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
Donkey anti-Rabbit IgG whole molecule
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
Rabbit IgG whole molecule
2 mg
Secondary
Rabbit
ELISA, Immunohistochemistry, Western Blot
Donkey anti-rabbit IgG recognizes rabbit IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography.
In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Donkey
This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities.
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal