Learn More
Donkey anti-Rat IgG whole molecule, Biotin, Polyclonal, Secondary Antibody, Abnova™
Manufacturer:  Abnova Corporation PAB10854
Catalog No. 89-137-388
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
Donkey anti-Rat IgG whole molecule
Biotin
The optimal working dilution should be determined by the end user.
Lyophilized
Rat IgG whole molecule
2 mg
Secondary
Rat
ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
Donkey anti-rat IgG recognizes rat IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Biotin.
Lyophilized from 10mM potassium phosphate, 75mM NaCl, 50% (v/v) glycerol, pH 7.2 (0.01% (w/v) sodium azide, 10mg/mL BSA (ImmunoglobulLyophilized from and Protease free))
Donkey
This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities.
Store at 4°C on dry atmosphere. After reconstitution with 1mL of deionized water, store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal