Learn More
Donkey anti-Sheep IgG whole molecule, AP, Polyclonal, Secondary Antibody, Abnova™
Manufacturer:  Abnova Corporation PAB10895
Catalog No. 89-137-427
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
Donkey anti-Sheep IgG whole molecule
AP
The optimal working dilution should be determined by the end user.
Liquid
Sheep IgG whole molecule
1 mg
Secondary
Sheep
Dot Blot, ELISA, Immunohistochemistry, Western Blot
Donkey anti-sheep IgG recognizes sheep IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Alkaline Phosphatase.
In 50mM Tris-Cl, 150mM NaCl, 1mM MgCl, 0.1mM ZnCl2, 50% (v/v) glycerol, pH 8.0 (0.01% sodium azide)
Donkey
This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities.
Store at 4°C. Do not Freeze.
Polyclonal