Learn More
Donkey anti-Sheep IgG whole molecule,Polyclonal Secondary Antibody, Abnova™
Manufacturer:  Abnova Corporation PAB9448
Catalog No. 89-139-100
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
Donkey anti-Sheep IgG whole molecule
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
Sheep IgG whole molecule
1 mg
Secondary
No reaction was observed against anti-Pepsin or anti-donkey IgG F(c).
ELISA, Immunohistochemistry, Western Blot
Donkey anti-sheep IgG recognizes sheep IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography.
In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Donkey
This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities, pepsin digestion and chromatographic separation.
Store at 4°C. Do not Freeze.
Polyclonal
Sheep