Learn More
Donkey IgG, F(ab)2, Fluorescein, Isotype control, Novus Biologicals™
Donkey Isotype Control
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP196843
Catalog No. NBP196843
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
IgG F(ab)2
Fluorescein
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: 10 mg/mL BSA - Immunoglobulin and Protease free. with 0.01% Thimerosal
Donkey
Ion exchange chromatography
RUO
Primary
Reconstitute with 1.0 ml deionized water (or equivalent) to desired concentration.
LYOPH
Restore with 1.0mL of deionized water (or equivalent).
Conjugated
IgG2
1 mg
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Fluorescein, anti-Donkey IgG, anti-Donkey IgG F(ab)2 and anti-Donkey Serum. No reaction was observed against anti-Donkey IgG F(ab)2 or anti-Pepsin.
Donkey