Learn More
Donkey IgG, F(ab)2, HRP, Isotype control, Novus Biologicals™
Donkey Isotype Control
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP197129
Catalog No. NBP197129
Add to cart
Description
Description
IgG F(ab)2 antibody specifically detects IgG F(ab)2 in Donkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
Specifications
IgG F(ab)2
HRP
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: 10 mg/mL BSA - Immunoglobulin and Protease free. with 0.01% Gentamicin Sulfate
Donkey
Ion exchange chromatography
RUO
Primary
Reconstitute with 1.0 ml deionized water (or equivalent) to desired concentration.
LYOPH
Restore with 1.0mL of deionized water (or equivalent).
Conjugated
IgG2
1 mg
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Peroxidase, anti-Donkey IgG, anti-Donkey IgG F(ab)2 and anti-Donkey Serum. No reaction was observed against anti-Donkey IgG F(c) or anti-Pepsin.
Donkey