Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BINDER™ Door Gasket

Silicone-free door gasket made of FKM.
$825.20 - $1064.25
Specifications
Description | Door Gasket |
---|---|
Material | Plastic |
Weight (English) | 2.20 lb. |
Weight (Metric) | 1 kg |
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
01-672-884
![]() |
BINDER™
80122333 |
Units M 260 |
Each for $1,064.25
|
|
|||||
01-672-885
![]() |
BINDER™
80122332 |
Units M 115 |
Each for $881.25
|
|
|||||
01-672-886
![]() |
BINDER™
80122331 |
Units M 56 |
Each for $825.20
|
|
|||||
Description
- Silicone-free
- Made of FKM
- For use with various BINDER™ Unit M lines
Specifications
Door Gasket | |
Plastic | |
2.20 lb. | |
1 kg |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title