Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dopamine D1R/DRD1 Mouse anti-Human, Alexa Fluor 350, Clone: 887438, R&D Systems

Mouse MonoclonalAntibody

Manufacturer:  R&D Systems FAB8276U100UG

Catalog No. FAB8276U100


Add to cart
This item is not returnable. View return policy

Description

Description

Dopamine D1R/DRD1 Monoclonal antibody specifically detects Dopamine D1R/DRD1 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Dopamine D1R/DRD1
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
P21728
DRD1
NS0 mouse myeloma cell line transfected with human Dopamine D1 R/DRD1 Met1-Thr446 Accession # P21728
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Stains human Dopamine D1 R/DRD1 transfected cells but not irrelevant transfectants in flow cytometry.
Flow Cytometry
887438
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
D(1A) dopamine receptor, D1DR, DADR, Dopamine D1 R, Dopamine D1 receptor, dopamine receptor D1, DRD1, DRD1A
Mouse
IgG2a
RUO
Primary
1812
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)