Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dopamine D3R/DRD3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen Dopamine D3R/DRD3
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB092587
View Documents
Novus Biologicals
NBP303577100UL
100 μg Each for $320.71
Add to cart
 
NB092588
View Documents
Novus Biologicals
NBP30357720UL
20 μg Each for $138.37
Add to cart
 
Description

Description

Dopamine D3R/DRD3 Polyclonal antibody specifically detects Dopamine D3R/DRD3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications

Specifications

Dopamine D3R/DRD3
Unconjugated
Rabbit
Human, Mouse, Rat
D(3) dopamine receptor, D3DR, Dopamine D3 receptor, dopamine receptor D3, essential tremor 1, ETM1, FET1, MGC149204, MGC149205
Recombinant fusion protein containing a sequence corresponding to amino acids 213-329 of human Dopamine D3R/DRD3 (NP_000787.2).
Polyclonal
Affinity purified
Immunofluorescence, Immunohistochemistry, Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
1814
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit