Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dopamine Transporter Rabbit anti-Human, Mouse, Rat, FITC, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc DAT1FITC
Description
Dopamine Transporter Polyclonal antibody specifically detects Dopamine Transporter in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western BlotSpecifications
Dopamine Transporter | |
Polyclonal | |
FITC | |
Slc6a3 | |
DA transporter; Dat; Dat1; dopamine transporter; dopamine transporter 1; PKDYS; SLC6A3; Sodium-dependent dopamine transporter; solute carrier family 6 (neurotransmitter transporter), member 3; solute carrier family 6 (neurotransmitter transporter, dopamine), member 3; solute carrier family 6 member 3; solute carrier family 6, member 3 | |
Synthetic peptide taken within amino acid region 550-600 on SLC6A3 protein. | |
200 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
P23977, Q01959, Q61327 | |
Rabbit | |
Affinity chromatography | |
RUO | |
13162, 24898, 6531 | |
-20° C, store in dark | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction