Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DOPEY1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | DOPEY1 |
---|---|
Concentration | 0.36 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-4962
![]() ![]() |
Proteintech Group Inc
255691AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene belongs to the dopey family. It is a transporter protein, possibly involved in protein traffic between late Golgi and early endosomes. This gene plays a potential role in functional brain alterations and in the pathogenesis of mental retardation in Down syndrome, it overexpression in the brain regions, that are altered in Down syndrome patients and involved in learning and memory processes.Specifications
DOPEY1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q5JWR5 | |
23033 | |
DOPEY1 Fusion Protein Ag22244 | |
Primary | |
-20°C | |
DOPEY1 |
0.36 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
dopey family member 1, DOPEY1, KIAA1117, Protein dopey 1 | |
DOPEY1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title