Learn More
Selleck Chemical LLCSupplier Diversity Partner Dorzolamide HCl 25mg 130693-82-2 MK-507 (L-671152) HCl  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S137525MG
Dorzolamide (MK-507, L-671152) HCl is a water-soluble, potent inhibitor of human carbonic anhydrase II and IV with Ki of 1.9 nM and 31 nM, respectively, used as anti-glaucoma agent.
Catalog No. 50-702-6