Learn More
Selleck Chemical LLC
Dorzolamide HCl 100mg 130693-82-2 MK-507 (L-671152) HCl
Manufacturer: Selleck Chemical LLC S1375100MG
Dorzolamide (MK-507, L-671152) HCl is a water-soluble, potent inhibitor of human carbonic anhydrase II and IV with Ki of 1.9 nM and 31 nM, respectively, used as anti-glaucoma agent.