Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Dosys™ 162 Basic Laboratory Syringes

Two-ring handled syringes with Luer Lock nozzles include a PVC protection sleeve and operating instructions.

Manufacturer:  DWK Life Sciences W852004

 View more versions of this product

Catalog No. 02-915-027


Due to product restrictions, please to purchase or view availability for this product.


Description

Description

  • Fill-by-fill (automatic) through needle or cannula
  • Supplied without valve system
  • Fast and reliable volume setting
  • Excellent reproducibility
  • Plunger tightness without O-ring
  • High chemical resistance
  • Fully autoclavable at 121°C
Specifications

Specifications

1 EA
Dosys 162
Syringes
Yes
PVC protection sleeve
Luer Lock nozzle
One
0.5–5mL
2-Ring handle
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit