Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ DOT Vehicle Placards: 1.6 EXPLOSIVE N 1

Comply with DOT regulations by marking delivery vehicles carrying dangerous or hazardous materials. Brady™ DOT Vehicle Placards are available in a variety of materials.

Manufacturer:  Brady™ 63369

 View more versions of this product

Catalog No. 19-111-296


  Check Availability
Add to cart

Specifications

Description

  • Available in three different materials Pressure Sensitive Vinyl, Polycoated Tagstock and Premium Fiberglass
Description & Specifications

Specifications

Non-adhesive
1.6 EXPLOSIVE N 1
Diamond
0.0984 in
Vehicle Placard
Black,Orange
Material Handling, Shipping & Receiving Signs & Labels
W x H: 10.75 x 10.75 in.
Fiberglass
1/ea
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit