Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ DOT Vehicle Placards: DANGEROUS
Comply with DOT regulations by marking delivery vehicles carrying dangerous or hazardous materials. Brady™ DOT Vehicle Placards are available in a variety of materials.
Supplier: Brady™ 76259
Description
- Available in three different materials Pressure Sensitive Vinyl, Polycoated Tagstock and Premium Fiberglass
Specifications
Non-adhesive | |
Fiberglass (B-120) | |
DANGEROUS | |
1 Ea. | |
10.75 x 10.75 in. |
Black, Red, White | |
Vehicle Placard | |
Material Handling, Shipping & Receiving Signs & Labels | |
Diamond | |
0.0984 in. |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction