Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ DOT Vehicle Placards: EXPLOSIVE 1.3C
Comply with DOT regulations by marking delivery vehicles carrying dangerous or hazardous materials. Brady™ DOT Vehicle Placards are available in a variety of materials.
Specifications
Adhesive Type | Non-adhesive |
---|---|
Color | Black, Orange |
Product Type | Vehicle Placard |
Display Text | EXPLOSIVE 1.3C |
For Use With (Application) | Material Handling, Shipping & Receiving Signs & Labels |
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
19-111-282
|
Brady™
63354 |
Fiberglass | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
19-111-311
|
Brady™
63389 |
Polycoated Tagstock | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Available in two different materials Polycoated Tagstock, Pressure Sensitive Vinyl
Specifications
Non-adhesive | |
Vehicle Placard | |
Material Handling, Shipping & Receiving Signs & Labels | |
Diamond |
Black, Orange | |
EXPLOSIVE 1.3C | |
1 Ea. | |
10.75 x 10.75 in. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title