Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ DOT Vehicle Placards: EXPLOSIVE 1.3F

Comply with DOT regulations by marking delivery vehicles carrying dangerous or hazardous materials. Brady™ DOT Vehicle Placards are available in a variety of materials.

$4.05 - $18.25

Specifications

Size (English) W x H: 10.75 x 10.75 in.
Product Type Vehicle Placard
Display Text EXPLOSIVE 1.3F
For Use With (Application) Material Handling, Shipping & Receiving Signs & Labels
Color Black, Orange
View More Specs

Products 3
Catalog Number Mfr. No. Material Price Quantity & Availability  
Catalog Number Mfr. No. Material Price Quantity & Availability  
19-111-253
View Documents
Brady™
63320
Pressure Sensitive Vinyl Each for $7.40
Add to cart
 
19-111-283
View Documents
Brady™
63355
Fiberglass Each for $18.25
Add to cart
 
19-111-312
View Documents
Brady™
63390
Polycoated Tagstock Each for $4.05
Add to cart
 
Description

Description

  • Available in three different materials Pressure Sensitive Vinyl, Polycoated Tagstock and Premium Fiberglass
Specifications

Specifications

W x H: 10.75 x 10.75 in.
EXPLOSIVE 1.3F
Black, Orange
Vehicle Placard
Material Handling, Shipping & Receiving Signs & Labels
Diamond
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit