Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ DOT Vehicle Placards: EXPLOSIVE 1.4C
Comply with DOT regulations by marking delivery vehicles carrying dangerous or hazardous materials. Brady™ DOT Vehicle Placards are available in a variety of materials.
Specifications
Color | Black, Orange |
---|---|
Product Type | Vehicle Placard |
Display Text | EXPLOSIVE 1.4C |
For Use With (Application) | Material Handling, Shipping & Receiving Signs & Labels |
Quantity | 1 Ea. |
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
19-111-260
|
Brady™
63327 |
Pressure Sensitive Vinyl (B-946) | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
19-111-290
|
Brady™
63362 |
Fiberglass (B-120) | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Available in two different materials Pressure Sensitive Vinyl, Premium Fiberglass
Specifications
Black, Orange | |
EXPLOSIVE 1.4C | |
1 Ea. | |
10.75 x 10.75 in. |
Vehicle Placard | |
Material Handling, Shipping & Receiving Signs & Labels | |
Diamond |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title