Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ DOT Vehicle Placards: EXPLOSIVE 1.4E

Comply with DOT regulations by marking delivery vehicles carrying dangerous or hazardous materials. Brady™ DOT Vehicle Placards are available in a variety of materials.

$4.05 - $18.25

Specifications

Size (English) W x H: 10.75 x 10.75 in.
Product Type Vehicle Placard
Display Text EXPLOSIVE 1.4E
For Use With (Application) Material Handling, Shipping & Receiving Signs & Labels
Color Black, Orange
View More Specs

Products 2
Catalog Number Mfr. No. Material Price Quantity & Availability  
Catalog Number Mfr. No. Material Price Quantity & Availability  
19-111-292
View Documents
Brady™
63364
Fiberglass
Each for $18.25
Add to cart
 
19-111-318
View Documents
Brady™
63399
Polycoated Tagstock
Each for $4.05
Add to cart
 
Description

Description

  • Available in two different materials Premium Fiberglass, Polycoated Tagstock
Specifications

Specifications

W x H: 10.75 x 10.75 in.
EXPLOSIVE 1.4E
Black, Orange
Vehicle Placard
Material Handling, Shipping & Receiving Signs & Labels
Diamond
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit